Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA28 Activator gamma Subunit/PSME3 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300356PE
Description
PA28 Activator gamma Subunit/PSME3 Polyclonal antibody specifically detects PA28 Activator gamma Subunit/PSME3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
PA28 Activator gamma Subunit/PSME3 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
11S regulator complex gamma subunit, 11S regulator complex subunit gamma, Activator of multicatalytic protease subunit 3, Ki, Ki antigen, PA28g, PA28gamma, PA28-gamma, PA28GKi nuclear autoantigen, proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki), Proteasome activator 28 subunit gamma, proteasome activator 28-gamma, proteasome activator complex subunit 3, REG-gamma | |
Produced in rabbits immunized with purified, recombinant Human PA28 Activator gamma Subunit/PSME3 (Accession#: P61289-1; Met1-Tyr254) | |
0.1 mL | |
Core ESC Like Genes, Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
10197 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction