Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA2G4/EBP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP190369
Description
PA2G4/EBP1 Polyclonal specifically detects PA2G4/EBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PA2G4/EBP1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, hG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PA2G4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KSDQDYILKEGDLVKIDLGVHVDGFIANVAHTFVVDVAQGTQVTGRKADVIKAAHLCAEAALRLVKPGNQNTQVTEAWNKVAHSFNCTPIEGMLSHQLKQHVIDGEKTIIQNPTDQQKKDHEKAEFEVHEVYAVDV | |
0.1 mL | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
5036 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction