Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA2G4/EBP1 Antibody (OTI1D3), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273213FR
Description
PA2G4/EBP1 Monoclonal specifically detects PA2G4/EBP1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PA2G4/EBP1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PA2G4 | |
Full length recombinant protein of human PA2G4/EBP1 (NP_006182) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1D3 | |
Western Blot | |
Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, hG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
5036.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction