Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PA2G4/EBP1 Antibody (SR1830), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32216450UL
Description
PA2G4/EBP1 Monoclonal antibody specifically detects PA2G4/EBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
PA2G4/EBP1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1830 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:30, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:40 | |
Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, hG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4 | |
A synthesized peptide derived from human PA2G4/EBP1 (Uniprot #: Q9UQ80) | |
50 μg | |
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
5036 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction