Learn More
PA2G4 Mouse anti-Human, Mouse, Rat, Clone: 3G10D9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | PA2G4 |
---|---|
Clone | 3G10D9 |
Concentration | 1.04 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6494
![]() ![]() |
Proteintech Group Inc
660551IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a mouse mono-clonal antibody raised against full length PA2G4 of human origin.
This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.Specifications
PA2G4 | |
1.04 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EBP1, ErbB3 binding protein 1, HG4 1, p38 2G4, PA2G4 | |
Pa2g4 | |
IgG2a | |
Protein A | |
Antibody |
3G10D9 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P50580, Q6AYD3, Q9UQ80 | |
18813, 288778, 5036 | |
PA2G4 Fusion Protein Ag7850 | |
Primary | |
-20°C | |
PA2G4 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.