Learn More
Motic™ PA43 BIO Phase Trinocular Routine Lab Microscope Package
The PA43 BIO models have been designed for research use in hospitals and life science laboratories. This package includes the PA43 BIO Trinocular Routine Lab Microscope with 0.5X C-mount camera adapter (PA53 Series), Moticam and Motic™ Analysis Basic 64-bit.
$7821.12
Specifications
Magnification Power 10X
Objective Types UC Plan 4X, 100X and UC PH Plan 10X, 40X
Head Style Trinocular
Lighting Type Kohler Illumination
Eyepiece WF 10X/22 mm
View More Specs
Products 1
Catalog Number Mfr. No. Camera Type Price Quantity & Availability  
Catalog Number Mfr. No. Camera Type Price Quantity & Availability  
01-257-437
View Documents
Motic™
PA43BIOPH1S6
6 MP Digital
Each of 1 for $7,821.12
Only null left
Add to Cart
Description
Description
  • Scalable Infinity-Corrected Optical System (CCIS™)
  • Full power transmitted Köhler illumination
  • Phase contrast methods for various microbiology applications
  • Darkfield accessories for blood cell studies for natural therapy
  • Polarizing contrast accessories for protein crystal, tissue, gout and kidney stone observations
  • Environmentally friendly 'ECO' function
  • Consistent, even illumination with IL Function
  • Rackless stage
  • Diopter Adjustment: ±5
Specifications
Specifications
10X
Trinocular
WF 10X/22 mm
UC Plan 4X, 100X and UC PH Plan 10X, 40X
Kohler Illumination
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.