Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZDHHC6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113325
Description
ZDHHC6 Polyclonal Antibody for Western Blot, ELISA
Endoplasmic reticulum palmitoyl acyltransferase that mediates palmitoylation of proteins such as AMFR, CALX, ITPR1 and TFRC (PubMed:22314232, PubMed:22728137, PubMed:25368151, PubMed:28826475). Palmitoylates calnexin (CALX), which is required for its association with the ribosome-translocon complex and efficient folding of glycosylated proteins (PubMed:22314232). Mediates palmitoylation of AMFR, promoting AMFR distribution to the peripheral endoplasmic reticulum (PubMed:22728137). Together with SELENOK, palmitoylates ITPR1 in immune cells, leading to regulate ITPR1 stability and function (PubMed:25368151). [UniProt].
Specifications
ZDHHC6 | |
Polyclonal | |
Unconjugated | |
ZDHHC6 | |
2400007G07Rik; 5930409M18Rik; AI462345; C77369; DHHC-6; H4 homolog; palmitoyltransferase ZDHHC6; probable palmitoyltransferase ZDHHC6; transmembrane protein H4; ZDHHC6; Zinc finger DHHC domain-containing protein 6; zinc finger DHHC-type containing 6; zinc finger protein 376; zinc finger, DHHC domain containing 6; zinc finger, DHHC-type containing 6; ZNF376 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64429, 66980 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CPV7, Q9H6R6 | |
ZDHHC6 | |
Recombinant Human Palmitoyltransferase ZDHHC6 protein (227-409AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction