Learn More
Invitrogen™ IPP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114035
Description
Immunogen sequence: EMQGLIYVIG GISNEGIELR SFEVYDPLSK RWSPLPPMGT RRAYLGVAAL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Zebrafish: 79%.
The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 50 amino acid repeat which interacts with actin. Transcript variants have been described but their full-length nature has not been determined.
Specifications
IPP | |
Polyclonal | |
Unconjugated | |
IPP | |
Actin-binding protein IPP; IAP promoted placental; intracisternal A particle-promoted polypeptide; IPP; kelch-like family member 27; kelch-like protein 27; KLHL27 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3652 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y573 | |
IPP | |
Synthetic peptide directed towards the C terminal region of human IPP (aa 434-483). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.