Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GFI1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117398
Description
Centrifuge briefly prior to opening.
GFI1 is a nuclear zinc finger (ZF) transcriptional repressor that plays an important role in hematopoiesis and inner ear development, and has been implicated in lymphomagenesis. The Gfi1 protein expression is regulated by the ubiquitin-proteasome system. It is reported that ubiquitin ligase Triad1 interacts with the DNA-binding domain of Gfi-1. Gfi-1 represses transcription by directly binding to the consensus DNA sequence in the promoters of its target genes. Deficiency of Gfi-1 causes neutropenia and an accumulation of granulomonocytic precursor cells that is reminiscent of a myelodysplastic syndrome in mice.
Specifications
GFI1 | |
Polyclonal | |
Unconjugated | |
GFI1 | |
AW495828; FLJ94509; Gfi1; GFI-1; GFI1A; growth factor independence-1; growth factor independent 1; growth factor independent 1 transcription repressor; growth factor independent 1 transcriptional repressor; growth factor independent protein 1; Growth factor independent-1; Pal1; Pal-1; SCN2; Zinc finger protein 163; zinc finger protein Gfi-1; ZNF163 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2672 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.27 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q99684 | |
GFI1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Human GFI1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction