Learn More
Motic™ PA53 MET Trinocular Upright Materials Microscope
Shop All Motic Group Products
Click to view available options
Lighting Type:
Halogen
Reflected Illumination
Reflected and Transmitted Illumination
Model:
PA53 MET
PA53 MET BD
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Superior optical performance with newly designed Semi-Apochromatic objectives
- Interference Contrast (DIC) concept incident illumination
- Established contrast methods
- Multiple degrees of motorization in X, Y, and Z
- Harmonized Brightfield/Darkfield switch
- Transmitted Light: LB daylight filter (Halogen lamp)
- Diopter adjustable
Specifications
Specifications
| Magnification Power | 10X |
| Objective Types | S-APO Plan 5X, 10X, 20X, 50X |
| Model | PA53 MET |
| Head Style | Trinocular |
| Lighting Type | Halogen |
| Eyepiece | WF 10X/25 mm |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov