Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAAF1 Antibody (OTI1F11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273214
Description
PAAF1 Monoclonal specifically detects PAAF1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PAAF1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:400-4000, Immunocytochemistry/Immunofluorescence 1:100 | |
FLJ11848, PAAF, proteasomal ATPase-associated factor 1, Protein G-16, Rpn14, WD repeat domain 71, WD repeat-containing protein 71, WDR71 | |
Mouse | |
42 kDa | |
100 μg | |
80227 | |
Human, Mouse | |
Lyophilized |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1F11 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PAAF1 | |
Full length human recombinant protein of human PAAF1(NP_079431) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction