Learn More
PAAF1 Monoclonal Antibody (OTI1F11)

Description
PAAF1 Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | PAAF1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1F11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PAAF1 |
| Gene Accession No. | Q9BRP4 |
| Gene Alias | PAAF; PAAF1; proteasomal ATPase associated factor 1; proteasomal ATPase-associated factor 1; Protein G-16; Rpn14; WD repeat domain 71; WD repeat-containing protein 71; WDR71 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.