Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PABP Antibody (10E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 8 publications
Supplier: Novus Biologicals NB1206125SS
Description
PABP Monoclonal specifically detects PABP in Human, Bovine, Canine, Primate, Xenopus, Drosophila (Negative), Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray, CyTOF-ready.Specifications
PABP | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000, Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:10-1:500, Microarray, CyTOF-ready | |
PAB1polyadenylate-binding protein 1, PABP, PABP-1, PABP1poly(A) binding protein, cytoplasmic 2, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 1, Poly(A)-binding protein 1, poly(A)-binding protein, cytoplasmic 2 | |
Mouse | |
69 kDa | |
0.025 mL | |
Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
26986 | |
Human, Bovine, Canine, Primate, Xenopus species, Drosophila (Negative), Mouse (Negative) | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
100000000000 | |
Unconjugated | |
P11940 | |
PABPC1 | |
Recombinant human PABP protein. [Uniprot# P11940] | |
Protein G purified | |
RUO | |
Primary | |
Human, Monkey, Bovine, Dog, and Hamster and Xenopus laevis. Does not cross-react with Drosophilia melanogaster or Mouse. Not yet tested in other species. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction