Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABP Monoclonal Antibody (10E10)
GREENER_CHOICE

Catalog No. PIMA546551
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546551 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546551 Supplier Invitrogen™ Supplier No. MA546551
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Western blot detects a band of approximately 70 kDa (predicted molecular weight: 71 kDa).

The poly(A)-binding protein (PABP), which is found complexed to the 3-prime poly(A) tail of eukaryotic mRNA, is required for poly(A) shortening and translation initiation. In humans, the PABPs comprise a small nuclear isoform and a conserved gene family that displays at least 3 functional proteins: PABP1 (PABPC1), inducible PABP (iPABP, or PABPC4), and PABP3 (PABPC3). In addition, there are at least 4 pseudogenes, PABPCP1 to PABPCP4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABP
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 10E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PABPC1
Gene Accession No. P11940, P20965
Gene Alias Cytoplasmic poly(A)-binding protein 1-A; ePAB; hypothetical protein; PAB1; pabp; PABP 1; Pabp1; PABP-1; PABP-1A; PABP-1-A; PABP-A; Pabpc1; pabpc1.L; pabpc1.S; pabpc1a; pabpc1-a; pabpc1a protein; pabpc1-b; pabpc2; PabpI; PABPL1; poly A binding protein 1; poly A binding protein, cytoplasmic 1 a; poly(A) binding protein cytoplasmic 1; poly(A) binding protein, cytoplasmic 1; poly(A) binding protein, cytoplasmic 1 L homeolog; poly(A) binding protein, cytoplasmic 1 S homeolog; poly(A) binding protein, cytoplasmic 1a; poly(A) binding protein, cytoplasmic 2; poly(A)-binding protein 1; poly(A)-binding protein 1A; poly(A)-binding protein 1-A; polyadenylate-binding protein 1; Polyadenylate-binding protein 1A; Polyadenylate-binding protein 1-A; polyadenylate-binding protein 1A; poly(A) binding protein, cytoplasmic 1a; wu:fb16a02; wu:fi19b08; wu:fj12d09; wu:fj61f06; XELAEV_18033915mg; xPABP1-A; zgc:109879
Gene Symbols PABPC1, pabpc1.S
Host Species Mouse
Immunogen Human recombinant PABP expressed from its 1.85 kbp cDNA, NcoI to SspI.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100342434, 100757538, 26986, 379896, 430997
Target Species Human, Rabbit, Hamster, Chicken, Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.