Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABPC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566900
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566900 Supplier Invitrogen™ Supplier No. PA566900
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QTQNRAAYYPP SQIAQLRPSP RWTAQG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

PABPC1b encodes a poly(A) binding protein The protein shuttles between the nucleus and cytoplasm and binds to the 3' poly(A) tail of eukaryotic messenger RNAs via RNA-recognition motifs The binding of this protein to poly(A) promotes ribosome recruitment and translation initiation; it is also required for poly(A) shortening which is the first step in mRNA decay The gene is part of a small gene family including three protein-coding genes and several pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABPC1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PABPC1
Gene Accession No. P11940, P29466, P48729, Q8N752
Gene Alias pabpc1; poly A binding protein, cytoplasmic 1 b; zgc:77608
Gene Symbols PABPC1
Host Species Rabbit
Immunogen Recombinant Human PABP. Recombinant protein control fragment (Product #RP-105419).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011, 1452, 26986, 834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.