Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PABPC1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PABPC1L |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PABPC1L Polyclonal specifically detects PABPC1L in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PABPC1L | |
Polyclonal | |
Rabbit | |
Human | |
80336 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ILTNQYMQRLSTMRTLSNPLLGSFQQPSSYFLPAMPQPPAQAAYYGCGPVTPTQP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
C20orf119, Chromosome 20 Open Reading Frame 119, DJ1069P2.3, Embryonic Poly(A) Binding Protein, EPAB, PABPC1L1, Poly(A) Binding Protein Cytoplasmic 1 Like, Poly(A) Binding Protein, Cytoplasmic 1-Like, Polyadenylate-Binding Protein 1-Like | |
PABPC1L | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title