Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABPC1L Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5121717
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121717 20 μL
PIPA5121716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121717 Supplier Invitrogen™ Supplier No. PA5121717
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PABPC1L is a protein coding gene. Gene Ontology (GO) annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABPC1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PABPC1L
Gene Accession No. Q4VXU2
Gene Alias 1810053B01Rik; C20orf119; dJ1069P2.3; embryonic poly(A) binding protein; embryonic poly(A)-binding protein; EPAB; LOW QUALITY PROTEIN: polyadenylate-binding protein 1-like; PABPC1L; PABPC1L1; poly(A) binding protein cytoplasmic 1 like; poly(A) binding protein, cytoplasmic 1-like; polyadenylate-binding protein 1-like; RGD1563142
Gene Symbols PABPC1L
Host Species Rabbit
Immunogen Recombinant Poly A Binding Protein Cytoplasmic 1 Like Protein (PABPC1L), Ala187-Lys324 (Accession Q4VXU2).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80336
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.