Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABPC1L2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541733 Supplier Invitrogen™ Supplier No. PA541733
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NGMFLNYRKI FVGRFKSHKE REAERGAWAR QSTSADVKDF EEDTDEEATL Sequence homology: Cow: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Rat: 77%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABPC1L2A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PABPC1L2A
Gene Accession No. Q5JQF8
Gene Alias PABPC1L2; Pabpc1l2a; PABPC1L2B; poly(A) binding protein cytoplasmic 1 like 2A; poly(A) binding protein, cytoplasmic 1-like 2A; poly(A) binding protein, cytoplasmic 1-like 2B; polyadenylate-binding protein 1-like 2; RBM32A; RBM32B; RGD1566367; RNA binding motif protein 32A; RNA binding motif protein 32B; RNA-binding motif protein 32; RNA-binding protein 32
Gene Symbols PABPC1L2A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PABPC1L2A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340529
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.