Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABPC3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522349

Catalog No. PIPA522349


Only null left
Add to Cart

Description

Description

Recommended positive controls: NT2D1, IMR32, Jurkat, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Messenger RNA stability and translation initiation are extensively under the control of poly(A)-binding proteins (PABP). See PABPC1 (MIM 604679) for background information.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PABPC3
Polyclonal
Unconjugated
PABPC3
4932702K14Rik; AI428050; PABP3; PABP-3; Pabpc3; Pabpc6; PABPL3; poly A binding protein, cytoplasmic 1 pseudogene; poly(A) binding protein cytoplasmic 3; poly(A) binding protein, cytoplasmic 3; poly(A) binding protein, cytoplasmic 6; poly(A)-binding protein 3; poly(A)-binding protein-like 3; polyadenylate-binding protein 3; RP11-165I9.5; testis PABP; Testis-specific poly(A)-binding protein; testis-specific poly(A)-binding protein (PABP); testis-specific poly(A)-binding protein 3; tPABP
Rabbit
Antigen affinity chromatography
RUO
5042
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.83 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9H361
PABPC3
Recombinant fragment corresponding to a region within amino acids 107 and 323 of PABPC3 (Uniprot ID#Q9H361).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.