Learn More
Invitrogen™ PABPC3 Polyclonal Antibody

Description
Recommended positive controls: Jurkat, Raji, K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PABPC3 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PABPC3 |
| Gene Accession No. | Q9H361 |
| Gene Alias | 4932702K14Rik; AI428050; PABP3; PABP-3; Pabpc3; Pabpc6; PABPL3; poly A binding protein, cytoplasmic 1 pseudogene; poly(A) binding protein cytoplasmic 3; poly(A) binding protein, cytoplasmic 3; poly(A) binding protein, cytoplasmic 6; poly(A)-binding protein 3; poly(A)-binding protein-like 3; polyadenylate-binding protein 3; RP11-165I9.5; testis PABP; Testis-specific poly(A)-binding protein; testis-specific poly(A)-binding protein (PABP); testis-specific poly(A)-binding protein 3; tPABP |
| Gene Symbols | PABPC3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.