Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PABPC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596483
Description
Immunogen sequence: VTEMNGRIVG SKPLYVALAQ RKEERKAHLT NQYMQRVAGM RALPANAILN QFQPAAGGYF VPAVPQAQGR PPYYTPNQLA QMRPNPRWQQ GGRPQGFQGM P; Positive Samples: HepG2, Jurkat, Mouse spleen, Mouse heart, Rat brain, Rat heart; Cellular Location: Cytoplasm.
Poly(A)-binding proteins (PABPs) bind to the poly(A) tail present at the 3-prime ends of most eukaryotic mRNAs. PABPC4 or IPABP (inducible PABP) was isolated as an activation-induced T-cell mRNA encoding a protein. Activation of T cells increased PABPC4 mRNA levels in T cells approximately 5-fold. PABPC4 contains 4 RNA-binding domains and proline-rich C terminus. PABPC4 is localized primarily to the cytoplasm. It is suggested that PABPC4 might be necessary for regulation of stability of labile mRNA species in activated T cells. PABPC4 was also identified as an antigen, APP1 (activated-platelet protein-1), expressed on thrombin-activated rabbit platelets. PABPC4 may also be involved in the regulation of protein translation in platelets and megakaryocytes or may participate in the binding or stabilization of polyadenylates in platelet dense granules. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
PABPC4 | |
Polyclonal | |
Unconjugated | |
PABPC4 | |
Activated-platelet protein 1; APP1; APP-1; Inducible poly(A)-binding protein; iPABP; PABP4; PABP-4; PABPC4; poly(A) binding protein cytoplasmic 4; poly(A) binding protein, cytoplasmic 4; poly(A) binding protein, cytoplasmic 4 (inducible form); poly(A)-binding protein 4; polyadenylate-binding protein 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
230721, 298510, 8761 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13310 | |
PABPC4 | |
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human PABPC4 (NP_0038101). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction