Learn More
Invitrogen™ PABPC5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531596
Description
Recommended positive controls: A431, HeLa, HepG2, U87-MG, mouse brain, rat brain. Predicted reactivity: Mouse (96%), Rat (97%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PABPC5 encodes a protein that binds to the polyA tail found at the 3' end of most eukaryotic mRNAs. It is thought to play a role in the regulation of mRNA metabolic processes in the cytoplasm. PABPC5 is located in a gene-poor region within the X-specific 13d-sY43 subinterval of the chromosome Xq21.3/Yp11.2 homology block. It is located close to translocation breakpoints associated with premature ovarian failure, and is therefore a potential candidate gene for this disorder.
Specifications
PABPC5 | |
Polyclonal | |
Unconjugated | |
PABPC5 | |
C820015E17; PABP5; PABP-5; Pabpc5; poly A binding protein, cytoplasmic 5; poly A binding protein, cytoplasmic 5-like; poly(A) binding protein cytoplasmic 5; poly(A) binding protein, cytoplasmic 5; polyadenylate-binding protein 5; polyadenylate-binding protein 5-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140886, 688518, 93728 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96DU9 | |
PABPC5 | |
Recombinant fragment corresponding to a region within amino acids 12 and 267 of Human PABP5. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.