Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABPC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551359 Supplier Invitrogen™ Supplier No. PA551359
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPNPAGKKKK YLKAALYVGD LDPDVTEDML YKKFRPAGPL RFTRICRDPV TRSPLGYGYV NFRFPADAEW ALNTMNFDLI NGKPFRLMWS QPDDRLRKSG VGNIFIKNLD KSIDNR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

PABPC5 encodes a protein that binds to the polyA tail found at the 3' end of most eukaryotic mRNAs. It is thought to play a role in the regulation of mRNA metabolic processes in the cytoplasm. PABPC5 is located in a gene-poor region within the X-specific 13d-sY43 subinterval of the chromosome Xq21.3/Yp11.2 homology block. It is located close to translocation breakpoints associated with premature ovarian failure, and is therefore a potential candidate gene for this disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABPC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PABPC5
Gene Accession No. Q96DU9
Gene Alias C820015E17; PABP5; PABP-5; Pabpc5; poly A binding protein, cytoplasmic 5; poly A binding protein, cytoplasmic 5-like; poly(A) binding protein cytoplasmic 5; poly(A) binding protein, cytoplasmic 5; polyadenylate-binding protein 5; polyadenylate-binding protein 5-like
Gene Symbols PABPC5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PABPC5. Recombinant protein control fragment (Product #RP-91974).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.