Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PABPN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PABPN1 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PABPN1 Polyclonal specifically detects PABPN1 in Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
PABPN1 | |
Polyclonal | |
Rabbit | |
Nuclear poly(A)-binding protein 1, OPMD, PAB2poly(A) binding protein II, PABII, PABP-2, PABP2polyadenylate-binding protein 2, poly(A) binding protein 2, poly(A) binding protein, nuclear 1, Poly(A)-binding protein 2, Poly(A)-binding protein II, poly(A)-binding protein, nuclear 1, Polyadenylate-binding nuclear protein 1 | |
PABPN1 | |
IgG | |
33 kDa |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
8106 | |
Synthetic peptides corresponding to PABPN1(poly(A) binding protein, nuclear 1) The peptide sequence was selected from the N terminal of PABPN1. Peptide sequence MAAAAAAAAAAGAAGGRGSGPGRRRHLVPGAGGEAGEGAPGGAGDYGNGL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title