Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PABPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596494 Supplier Invitrogen™ Supplier No. PA596494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAAAAAAA AGAAGGRGSG PGRRRHLVPG AGGEAGEGAP GGAGDYGNGL ESEELEPEEL LLEPEPEPEP EEEPPRPRAP PGAPGPGPGS GAPGSQEEEE; Positive Samples: B cells, HeLa, HepG2, U-87MG, Mouse brain, Mouse testis; Cellular Location: Cytoplasm, Nucleus.

This gene encodes an abundant nuclear protein that binds with high affinity to nascent poly(A) tails. The protein is required for progressive and efficient polymerization of poly(A) tails at the 3' ends of eukaryotic transcripts and controls the size of the poly(A) tail to about 250 nt. At steady-state, this protein is localized in the nucleus whereas a different poly(A) binding protein is localized in the cytoplasm. This gene contains a GCG trinucleotide repeat at the 5' end of the coding region, and expansion of this repeat from the normal 6 copies to 8-13 copies leads to autosomal dominant oculopharyngeal muscular dystrophy (OPMD) disease. Related pseudogenes have been identified on chromosomes 19 and X. Read-through transcription also exists between this gene and the neighboring upstream BCL2-like 2 (BCL2L2) gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PABPN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PABPN1
Gene Accession No. Q86U42, Q8CCS6
Gene Alias mPABII; Nuclear poly(A)-binding protein 1; OPMD; PAB2; PABII; PABP2; PABP-2; Pabp3; PABPN1; poly(A) binding protein 2; poly(A) binding protein II; poly(A) binding protein nuclear 1; poly(A) binding protein, nuclear 1; poly(A)-binding protein 2; poly(A)-binding protein II; polyA binding protein II; polyadenylate-binding nuclear protein 1; Polyadenylate-binding protein 2
Gene Symbols PABPN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PABPN1 (NP_0046341).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54196, 8106
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.