Learn More
Description
Combine Thermo Scientific™ μPAC™ Neo trapping columns with Thermo Scientific™ μPAC™ Neo low-load HPLC columns for optimal performance of bottom-up proteomics experiments.
Combine μPAC Neo low-load trapping columns with μPAC Neo low-load HPLC columns for optimal performance of bottom-up proteomics experiments. Specifically designed for single-cell proteomics applications, these trapping columns support the use of TMT-labeled samples, with up 10 ng of sample load.
Sample Preconcentration - Perform sample preconcentration in your preferred workflow, with both back flush and forward flush mobile phase direction capabilities.
Decreased Sample Loading Times - Shorten low-flow HPLC analysis run times by injecting your samples at 5-10 μL/min.
Sample Clean-up - Effectively remove salts, reagents, and contaminants that have no affinity for hydrophobic stationary phases from your samples, increasing your analytical column's lifetime and improving spectral quality and detection sensitivity.
Specifications
Specifications
| Flow Rate | 5 to 20 μL/min |
| Max. Pressure | 5800 psi (400 bar) |
| pH | 1.5 to 7 |
| Porosity | 59% |
| Quantity | 2 |
| Temperature | 40°C |
| Stationary Phase | C8 |
| Packing Material | Silicon Chip |
| Column Type | Micro Pillar Array |
| Endcapped | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.