Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ μPAC™ Trapping Column, 2/pk

Catalog No. 01258267 Shop All Thermo Scientific Products
Change view
Click to view available options
Pillar Length:
18 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pillar Length
01-258-267 18 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-258-267 Supplier Thermo Scientific™ Supplier No. COLTRPNANO16G1B2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform large volume (>5 μL) sample injections on capillary and nano-columns with μPAC™ Trapping Columns.

Perform large volume (>5 μL) sample injections on capillary and nano-columns with Thermo Scientific™ μPAC™ Trapping Columns with minimal impact on your total low-flow chromatography analysis time. Their C18 stationary phase support morphology is designed to perfectly match that of capillary and nano- μPAC HPLC columns, with carefully selected dimensions and surface chemistry.

Sample Preconcentration - Preconcentrate samples with very low peptide contents into a sub-microliter trapping volume.

Decreased Sample Loading Times - Depending on the flow rate of the analytical column used, a 10 to 50-fold reduction of the sample loading time can be achieved for high volume (5 to 20 μL) samples.

Sample Clean-up - Effectively remove salts, reagents, and other contaminants that have no affinity for hydrophobic bonded phases, increasing analytical column lifetime and improving spectral quality and detection sensitivity.

TRUSTED_SUSTAINABILITY

Specifications

Flow Rate 10 to 20 μL/min
Max. Pressure 350 bar (5076 psi)
pH 1.5 to 7
Pore Size 100 to 200 Å
Porosity 59%
Temperature 60°C
Stationary Phase C18
Packing Material Silicon Chip
Column Type Micro Pillar Array
Endcapped Yes
Bed Length 10 mm
Bed Width 2 mm
Interpillar Distance 2.5 μm
Pillar Diameter 5 μm
Pillar Length 18 μm
Product Type μPAC Trapping Columns
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.