Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAC2 Antibody (OTI1E8), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273215AF647
Description
PAC2 Monoclonal specifically detects PAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PAC2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PSMG2 | |
Full length recombinant protein of human PAC2 (NP_064617) produced in E.coli. | |
0.1 mL | |
56984.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1E8 | |
Western Blot, Immunohistochemistry | |
CLAST3HSPC260, HCCA3PAC-2, hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-susceptibility protein 3, HsT1707, likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3), MDS003, MGC15092, PAC2HDCMC29P, proteasome (prosome, macropain) assembly chaperone 2, proteasome assembling chaperone 2, proteasome assembly chaperone 2, TNFSF5IP1CD40 ligand-activated specific transcript 3, Tumor necrosis factor superfamily member 5-induced protein 1, tumor necrosis factor superfamily, member 5-induced protein 1, x 003 protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction