Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACAP Monoclonal Antibody (ZP006), Invitrogen™

Catalog No. PI396300
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI396300 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI396300 Supplier Thermo Scientific Supplier No. 396300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PACAP Monoclonal antibody specifically detects PACAP in Human, Rat samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACAP
Applications ELISA, Western Blot
Classification Monoclonal
Clone ZP006
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene ADCYAP1
Gene Accession No. P13589, P18509
Gene Alias ADCYAP1; adenylate cyclase activating polypeptide 1; adenylate cyclase activating polypeptide 1 (pituitary); HSPC190; MGC126852; OTTHUMP00000162201; PACAP; pacap 38; PACAP-27; PACAP-38; PACAP-related peptide; pERp1; pituitary adenylate cyclase activating polypeptide; pituitary adenylate cyclase activating polypeptide 1; pituitary adenylate cyclase-activating polypeptide; Pituitary adenylate cyclase-activating polypeptide 27; Pituitary adenylate cyclase-activating polypeptide 38; pituitary adenylate cyclase-activating polypeptide precursor (Pacap); prepro-PACAP; PRP-48
Gene Symbols ADCYAP1
Host Species Mouse
Immunogen Synthetic peptide derived from the C-terminal region of the human, mouse, and rat PACAP protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116, 24166
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.