Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACAP Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3213 Supplier Invitrogen™ Supplier No. PA3213
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA3-213 detects PACAP type 1A receptor in rat and mouse samples. PA3-213 has successfully been used in Western blot and immunohistochemical (paraffin) procedures. The PA3-213 immunogen is a synthetic peptide corresponding to the C-terminal of rat PAC1 (503-523) CLSKSSSQLRMSSLPADNLAT coupled to KLH.

This gene encodes type I adenylate cyclase activating polypeptide receptor, which is a membrane-associated protein and shares significant homology with members of the glucagon/secretin receptor family. This receptor mediates diverse biological actions of adenylate cyclase activating polypeptide 1 and is positively coupled to adenylate cyclase. Alternative splicing of two exons of this gene generates four major splice variants, but their full-length nature has not been determined. PACAP receptor type 1 expression has been documented in adipose, adrenal, bone, brain, colon, ganglion, heart, lung, ovary, pancreas, placenta, spleen, and uterus. ESTs have been isolated from brain and nerve libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACAP Receptor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.02% sodium azide
Gene ADCYAP1R1
Gene Accession No. P32215, P70205
Gene Alias 2900024I10Rik; ADCYAP receptor type I; ADCYAP1 R1; ADCYAP1R 1; Adcyap1r1; adenylate cyclase activating polypeptide 1 (pituitary) receptor type I; adenylate cyclase activating polypeptide 1 receptor 1; adenylate cyclase activating polypeptide 1 type 1 receptor; AI846590; alternatively spliced product of PACAP receptor; G-protein coupled receptor; PAC 1; PAC1; PAC1 receptor; Pac1 receptors; PAC1R; PAC1-R; PACAP R 1; PACAP Receptor; PACAP receptor 1; PACAP type I receptor; PACAP type IA receptor; PACAP/VIP receptors; PACAP1 R; PACAP1 receptor; PACAP1-R; PACAPR; PACAPR1; PACAP-R1; PACAP-R-1; PACAPR1A; PACAP-R1A; PACAP-R-1A; PACAPRI; pituitary adenylate cyclase activating polypeptide 1 receptor; pituitary adenylate cyclase activating polypeptide 1 receptor (1); pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop; pituitary adenylate cyclase activating polypeptide receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor hop 1 splice variant; pituitary adenylate cyclase-activating polypeptide type I receptor; pituitary adenylate cyclase-activating polypeptide type IA receptor; pvr1
Gene Symbols ADCYAP1R1
Host Species Rabbit
Immunogen C-terminal of rat PAC1 (503-523) CLSKSSSQLRMSSLPADNLAT coupled to KLH.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11517, 24167
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.