Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PACAP Receptor Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596229
Description
Positive Samples: HeLa, SH-SY5Y, U-87MG; Cellular Location: Cell membrane, Multi-pass membrane protein Immunogen sequence: CIFKKEQAMC LEKIQRANEL MGFNDSSPGC PGMWDNITCW KPAHVGEMVL VSCPELFRIF NPDQVWETET IGESDFGDSN SLDLSDMGVV SRNCTEDGWS EPFPHYFDAC GFDEYE.
This gene encodes type I adenylate cyclase activating polypeptide receptor, which is a membrane-associated protein and shares significant homology with members of the glucagon/secretin receptor family. This receptor mediates diverse biological actions of adenylate cyclase activating polypeptide 1 and is positively coupled to adenylate cyclase. Alternative splicing of two exons of this gene generates four major splice variants, but their full-length nature has not been determined. PACAP receptor type 1 expression has been documented in adipose, adrenal, bone, brain, colon, ganglion, heart, lung, ovary, pancreas, placenta, spleen, and uterus. ESTs have been isolated from brain and nerve libraries.
Specifications
PACAP Receptor | |
Polyclonal | |
Unconjugated | |
ADCYAP1R1 | |
2900024I10Rik; ADCYAP receptor type I; ADCYAP1 R1; ADCYAP1R 1; Adcyap1r1; adenylate cyclase activating polypeptide 1 (pituitary) receptor type I; adenylate cyclase activating polypeptide 1 receptor 1; adenylate cyclase activating polypeptide 1 type 1 receptor; AI846590; alternatively spliced product of PACAP receptor; G-protein coupled receptor; PAC 1; PAC1; PAC1 receptor; Pac1 receptors; PAC1R; PAC1-R; PACAP R 1; PACAP Receptor; PACAP receptor 1; PACAP type I receptor; PACAP type IA receptor; PACAP/VIP receptors; PACAP1 R; PACAP1 receptor; PACAP1-R; PACAPR; PACAPR1; PACAP-R1; PACAP-R-1; PACAPR1A; PACAP-R1A; PACAP-R-1A; PACAPRI; pituitary adenylate cyclase activating polypeptide 1 receptor; pituitary adenylate cyclase activating polypeptide 1 receptor (1); pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop; pituitary adenylate cyclase activating polypeptide receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor hop 1 splice variant; pituitary adenylate cyclase-activating polypeptide type I receptor; pituitary adenylate cyclase-activating polypeptide type IA receptor; pvr1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11517, 117, 24167 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.84 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P32215, P41586, P70205 | |
ADCYAP1R1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-140 of human ADCYAP1R1 (NP_0011092). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction