Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACAP Receptor Polyclonal Antibody
PACAP Receptor Polyclonal Antibody
Supplier: Thermo Scientific PA576696
Description
PACAP Receptor Polyclonal Antibody for Western Blot

Specifications
PACAP Receptor | |
Polyclonal | |
Unconjugated | |
ADCYAP1R1 | |
2900024I10Rik; ADCYAP receptor type I; ADCYAP1 R1; ADCYAP1R 1; Adcyap1r1; adenylate cyclase activating polypeptide 1 (pituitary) receptor type I; adenylate cyclase activating polypeptide 1 receptor 1; adenylate cyclase activating polypeptide 1 type 1 receptor; AI846590; alternatively spliced product of PACAP receptor; G-protein coupled receptor; PAC 1; PAC1; PAC1 receptor; Pac1 receptors; PAC1R; PAC1-R; PACAP R 1; PACAP Receptor; PACAP receptor 1; PACAP type I receptor; PACAP type IA receptor; PACAP/VIP receptors; PACAP1 R; PACAP1 receptor; PACAP1-R; PACAPR; PACAPR1; PACAP-R1; PACAP-R-1; PACAPR1A; PACAP-R1A; PACAP-R-1A; PACAPRI; pituitary adenylate cyclase activating polypeptide 1 receptor; pituitary adenylate cyclase activating polypeptide 1 receptor (1); pituitary adenylate cyclase activating polypeptide 1 receptor type I Hiphop; pituitary adenylate cyclase activating polypeptide receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor; pituitary adenylate cyclase-activating polypeptide type 1 receptor hop 1 splice variant; pituitary adenylate cyclase-activating polypeptide type I receptor; pituitary adenylate cyclase-activating polypeptide type IA receptor; pvr1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11517, 117, 24167 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P32215, P41586, P70205 | |
ADCYAP1R1 | |
Recombinant full length Human PACAP Receptor. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction