Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACE4/PCSK6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP314443
This item is not returnable.
View return policy
Description
PACE4/PCSK6 Polyclonal antibody specifically detects PACE4/PCSK6 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PACE4/PCSK6 | |
Polyclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5046 | |
Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.4.21, EC 3.4.21.61, EC 3.4.21.75, PACE4EC 3.4.21.-, Paired basic amino acid cleaving enzyme 4, paired basic amino acid cleaving system 4, proprotein convertase subtilisin/kexin type 6, SPC4subtilisin-like protease, Subtilisin/kexin-like protease PACE4, Subtilisin-like proprotein convertase 4 | |
Synthetic 14 amino acid peptide from Cys-rich motif (CRM) region of human PACE4/PCSK6. | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction