Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Pacific Orange™ and Pacific Blue™ Antibody Labeling Kits

Catalog No. p30013
Encompass_Preferred
Click to view available options
Excitation/Emission:
405/551 nm
410/455 nm
Color:
Pacific Blue
Pacific Orange

Antibody Labeling Kits specifically bind small amounts of antibody with violet light-excitable Pacific Orange or Pacific Blue dyes. Conveniently label up to 100 μg of antibody per reaction, with 5 reactions per kit.

Pacific Orange and Pacific Blue antibody labeling kits enable attachment of a violet light-excitable fluorescent label on antibodies (up to 100 μg per reaction). Each kit contains enough dye for five reactions.

A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Primary antibodies of the same isotype or derived from the same species can be multiplexed if they are directly labeled with compatible fluorophores. Using Pacific Blue and Pacific Orange dyes simultaneously allows you to excite both dyes at the same wavelength while their emissions are differentially distinguished.

Pacific Orange and Pacific Blue antibody labeling kits allow the labeling of small amounts of antibody with a violet light-excitable dye (up to 100 μg of antibody per reaction). Small amounts of other proteins (>40 kDa) can also be labeled using these kits.

Features of these antibody labeling kits
• Pacific Blue excitation and emission maxima: 410/455 nm
• Pacific Orange excitation and emission maxima: 405/551 nm
• Labeled proteins typically ready to use in 90 min. (∼15 min. hands-on time)
• Optimized for small-scale labeling of any protein >40 kDa
• Stabilizing proteins should be removed from the sample before labeling
• Purified using convenient spin filters
• Includes detailed instructions for determining degree of labeling (DOL)

These kits contain everything you need to perform five separate labeling reactions as well as to purify the resulting conjugates. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry, primary detection, ELISA, immunocytochemistry, indirect FISH, and more. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Labeling Kit
Quantity 5 Reactions kit
Content And Storage Contains 5 vials of Pacific Blue™ reactive dye, 1 vial sodium bicarbonate (84 mg), purification resin (10 mL), 5 spin columns, and and 5 collection tubes.

Store at 2-8°C, protected from light. Component A may be stored from -20°C to 8°C.
Detection Method Fluorescence
Excitation/Emission 410/455 nm
Labeling Scale 100 μg
Shipping Condition Room Temperature
Product Line Pacific Blue™
Chemical Reactivity Amine
Solubility DMSO (Dimethylsulfoxide)
Label or Dye Pacific Blue
Color Pacific Blue
Labeling Target Antibodies, Proteins
Label Type Pacific Dyes
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.