Learn More
Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls
Catalog No. p-4528050
Click to view available options
Type:
Fibrinogen High Control
Fibrinogen Low Control
This item is not returnable. View return policy
Catalog No. 23-033240
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Provide accurate, reproducible confirmation of coagulation test system performance with these coagulation controls.
Provide accurate, reproducible confirmation of coagulation test system performance with Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls. Controls can be run on most coagulation systems and are formulated to reflect expected patient results.
Coagulation and Heparin control plasmas are prepared from fresh pools of human plasma.
Coagulation Control Plasmas:
  • Normal control (Level 1) produces normal PT and aPTT times
  • Abnormal controls monitor therapeutic (Level 2) and highly elevated (Level 3) PT/aPTT times 
Heparin Control Plasmas:
  • Simulate low (Level 1) and elevated (Level 2) plasma levels of Heparin
  • Use to monitor and evaluate patients on Heparin therapy
TRUSTED_SUSTAINABILITY
Specifications
Type Fibrinogen High Control
Quantity 10 x 1 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.