Learn More
Thermo Scientific™ Pacific Hemostasis™ Coagulation Reference Plasmas
Description
Establish and check factor assay reference curves with Thermo Scientific™ Pacific Hemostasis™ Coagulation Reference Plasmas. Plasmas offer Universal (UCRP), Abnormal (ACRP) and Fibrinogen Reference Plasmas to meet the needs of low and high volume coagulation testing labs.
Universal Fibrinogen Reference Plasma is a normal human reference plasma. Analytes in this product contain clottable Protein C and Protein S. Abnormal Fibrinogen Reference Plasma is adjusted to produce coagulation factors in the abnormal range at approximately 50% of the UCRP product. Fibrinogen Reference Plasma has an assayed fibrinogen level for reference curve preparation.
UCRP and ACRP contain Factor I (Fibrinogen), Factor II (Prothrombin), Factor V, Factor VII, Factor VIII, Factor IX, Factor X, Factor XI, Factor XII, AT-III Chromogenic, vWF Antigen, Plasminogen, Protein C, Alpha-2-Antiplasmin and Protein S.
Specifications
Specifications
| Type | UCRP |
| Quantity | 10 X 1 mL/Pk. |