Learn More
Thermo Scientific™ Pacific Hemostasis™ Coagulation Reference Plasmas
Catalog No. p-4528059
Change view
Click to view available options
Type:
UCRP
Catalog No. Type
23-176172 UCRP
1 options
This item is not returnable. View return policy
Catalog No. 23-176172
This item is not returnable. View return policy
Establish and check factor assay reference curves with these coagulation reference plasmas.
Establish and check factor assay reference curves with Thermo Scientific™ Pacific Hemostasis™ Coagulation Reference Plasmas. Plasmas offer Universal (UCRP), Abnormal (ACRP) and Fibrinogen Reference Plasmas to meet the needs of low and high volume coagulation testing labs.
Universal Fibrinogen Reference Plasma is a normal human reference plasma. Analytes in this product contain clottable Protein C and Protein S. Abnormal Fibrinogen Reference Plasma is adjusted to produce coagulation factors in the abnormal range at approximately 50% of the UCRP product. Fibrinogen Reference Plasma has an assayed fibrinogen level for reference curve preparation.
UCRP and ACRP contain Factor I (Fibrinogen), Factor II (Prothrombin), Factor V, Factor VII, Factor VIII, Factor IX, Factor X, Factor XI, Factor XII, AT-III Chromogenic, vWF Antigen, Plasminogen, Protein C, Alpha-2-Antiplasmin and Protein S.
TRUSTED_SUSTAINABILITY
Specifications
Type UCRP
Quantity 10 X 1 mL/Pk.