Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Package A - Battery Test Chambers Safety Package for Aging Tests
Shop All BINDER Inc. Products

Click to view available options
Material:
Steel
Description
Aging tests:
These involve testing at a certain temperature without the battery load cycle. They are performed within a safe temperature range for the battery.
This safety package is ideal for tests that present a low potential hazard level and is exactly in line with EUCAR hazard classification 2.
Solution:
- Class 2 independent temperature safety device when temperature is set to 120°C
- Temperature range limited to 110°C on the controller
The packages can be adapted to a variety of BINDER models in order to meet test requirements relating to temperature range, climate range, and test area size.
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers |
| Material | Steel |
| Type | Option for BINDER Units |
| Features | Temperature range limitation |
| For Use With (Application) | Battery Testing |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction