Learn More
BINDER™ Package P - Battery Test Chambers Safety Package for Performance Tests

Description
Performance tests:
Various battery-specific parameters, such as the load state, are tested with overlapping temperature ranges. These tests are performed within a safe temperature range for the battery.
The equipment in this safety package is designed for tests on lithium-ion batteries and is exactly in line with EUCAR hazard classification 4.
Solution:
- Class 2 independent temperature safety device when temperature is set to 120°C
- Temperature range limited to 110°C on the controller
- Stainless-steel pressure relief flap installed in the middle on top of the unit
- Enhanced door-locking mechanism with reinforced brackets
- Inert gas connections
The packages can be adapted to a variety of BINDER models in order to meet test requirements relating to temperature range, climate range, and test area size.
Specifications
Specifications
For Use With (Equipment) | BINDER Chambers |
Material | Steel |
Type | Option for BINDER Units |
Features | Temperature range limitation, reversible pressure relief flap, door-locking mechanism, inert gas connections |
For Use With (Application) | Battery Testing |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.