Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Package P Plus - Battery Test Chambers Safety Package for Extended Performance Tests
SDP

Catalog No. 08112005 Shop All BINDER Inc. Products
Click to view available options
Material:
Steel

To conduct tests on lithium-ion batteries, BINDER offers temperature and climate chambers equipped with a range of safety packages that are aligned with EUCAR Hazard Standards, environmental requirements, or test types.

Performance tests:

Various battery-specific parameters, such as the load state, are tested with overlapping temperature ranges. These tests are performed within a safe temperature range for the battery.

The equipment in this safety package is designed for tests on lithium-ion batteries and is exactly in line with EUCAR hazard level 5.

Solution:

  • Class 2 independent temperature safety device when temperature is set to 120°C
  • Temperature range limited to 110°C on the controller
  • Stainless-steel pressure relief flap installed in the middle on top of the unit
  • Enhanced door-locking mechanism with reinforced brackets
  • Inert gas connections
  • CO2 fire suppression equipment

The packages can be adapted to a variety of BINDER models in order to meet test requirements relating to temperature range, climate range, and test area size.

Specifications

For Use With (Equipment) BINDER Chambers
Material Steel
Type Option for BINDER Units
Features Temperature range limitation, reversible pressure relief flap, door-locking mechanism, inert gas connections, temperature monitoring, warning indicator lamp, CO2 fire suppression equipment, manual activation
For Use With (Application) Battery Testing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.