Learn More
BINDER™ Package S - Battery Test Chambers Safety Package for Stress Tests
Shop All BINDER Inc. Products

Description
Stress tests:
Various battery-specific parameters, such as the load state, are tested with overlapping temperature ranges. These tests are performed at the limit of the safe temperature range for the battery. This category also includes tests with higher charging and discharging currents overlapping with constant or dynamic temperatures.
The extensive equipment in this safety package is designed for tests on lithium-ion batteries and is exactly in line with EUCAR hazard classification 6.
Solution:
- Class 2 independent temperature safety device when temperature is set to 120°C
- Temperature range limited to 110°C on the controller
- Stainless-steel reversible pressure relief flap installed in the middle on top of the unit
- Enhanced door-locking mechanism with reinforced brackets
- Monitoring of CO, H2, and O2 in testing area atmosphere
- N2 permanent inertization
- CO2 flushing device
The packages can be adapted to a variety of BINDER models in order to meet test requirements relating to temperature range, climate range, and test area size.
Specifications
Specifications
For Use With (Equipment) | BINDER Chambers |
Material | Steel |
Type | Option for BINDER Units |
Features | Temperature range limitation, reversible pressure relief flap, door-locking mechanism, gas detection, inertization equipment, warning indicator lamp, CO2 fire suppression equipment, manual activation |
For Use With (Application) | Battery Testing |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.