Learn More
Stoelting™ Packer Mosquito Hemostatic Forceps
SDP
Stoelting™ Packer Mosquito Hemostatic Forceps are used in many surgical procedures usually to control bleeding or the flow of liquids in tubing.
$325.50
Specifications
For Use With (Application) Neuroscience
Length (English) 5 in.
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-001-324
SDP
View Documents
Stoelting™
5211004
Straight, Flat
Each for $325.50
Only null left
Add to Cart
10-001-325
SDP
View Documents
Stoelting™
5211005
Curved, Flat
Each for $325.50
Only null left
Add to Cart
Description
Description
  • Scissor-like ring handles just like scissors
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
  • Delicate hemostats have thin jaws with fine serrations
Specifications
Specifications
Neuroscience
5 in.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.