Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACRG Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988960
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8960 Supplier Bioss Supplier No. BS9289R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PACRG is a protein that is conserved across metazoans. In vertebrates, this gene is linked in a head-to-head arrangement with the adjacent parkin gene, which is associated with autosomal recessive juvenile Parkinson′s disease. These genes are co-regulated in various tissues and they share a bi-directional promoter. Both genes are associated with susceptibility to leprosy. The parkin co-regulated gene protein forms a large molecular complex with chaperones, including heat shock proteins 70 and 90, and chaperonin components. This protein is also a component of Lewy bodies in Parkinson′s disease patients, and it suppresses unfolded Pael receptor-induced neuronal cell death.

Specifications

Antigen PACRG
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PACRG
Gene Accession No. Q96M98
Gene Alias 1700008H23Rik; BOS_10014; FLJ32724; GLUP; HAK005771; Hypertension-related protein 1-like protein; LOC100342049; Molecular chaperone/chaperonin-binding protein; PACRG; PARK2 coregulated; PARK2 co-regulated; PARK2 coregulated gene protein; PARK2CRG; parkin coregulated; Parkin coregulated gene protein; parkin co-regulated gene protein; Parkin coregulated gene protein homolog; RGD1561027; RP3-495O10.2; zgc:101786
Gene Symbols PACRG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PACRG.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135138
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.