Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572865
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572865 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572865 Supplier Thermo Scientific Supplier No. PA572865
Add to Cart
Add to Cart

PACS1 Polyclonal Antibody

PACS1 Polyclonal Antibody for Western Blot, IHC, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen PACS1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PACS1
Gene Accession No. O88588, Q6VY07, Q8K212
Gene Alias AI325977; cytosolic sorting protein PACS-1; KIAA1175; LOW QUALITY PROTEIN: phosphofurin acidic cluster sorting protein 1; phosphofurin acidic cluster sorting protein 1; MRD17; PACS1; PACS-1; Phosphofurin acidic cluster sorting protein 1
Gene Symbols PACS1
Host Species Rabbit
Immunogen PACS1 antibody for a 17 amino acid peptide near the center of human PACS1. The immunogen is within amino acids 430 - 480 of PACS1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107975, 171444, 55690
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.