Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PACS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145087
Description
Immunogen sequence: GSVDSKYSSS FLDSGWRDLF SRSEPPVSEQ LDVAGRVMQY VNGAATTHQL P Antigen sequence identity: Mouse 88%; Rat 88%.
PACS-1 (CRA_a) is a 963 amino acid rich regulatory protein involved in protein trafficking and belongs to the PACS family. A coat protein involved in controlling the correct subcellular localization of trans-Golgi network (TGN) membrane proteins that contain acidic cluster sorting motifs, PACS-1 is also known to be involved in HIV-1 Nef-mediated removal of MHC-I from the cell surface to the TGN, thus enabling HIV-1 to escape immune surveillance. PACS-1 also controls the endosome to Golgi trafficking of furin and mannose-6-phosphate receptor by connecting the acidic-cluster-containing cytoplasmic domain of these molecules with the adapter-protein complex-1 (AP-1) of endosomal clathrin coated membrane pits. Usually seen in golgi apparatus, trans-Golgi network and localized in the paranuclear region, it is ubiquitously expressed in most of the tissues.
Specifications
PACS1 | |
Polyclonal | |
Unconjugated | |
PACS1 | |
AI325977; cytosolic sorting protein PACS-1; KIAA1175; LOW QUALITY PROTEIN: phosphofurin acidic cluster sorting protein 1; phosphofurin acidic cluster sorting protein 1; MRD17; PACS1; PACS-1; Phosphofurin acidic cluster sorting protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55690 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6VY07 | |
PACS1 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 674-724. Recombinant protein control fragment (Product #a href=. | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction