Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523291
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523291 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523291 Supplier Invitrogen™ Supplier No. PA523291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence homology: Opossum/Canine (89%).

PACS-1 (CRA_a) is a 963 amino acid rich regulatory protein involved in protein trafficking and belongs to the PACS family. A coat protein involved in controlling the correct subcellular localization of trans-Golgi network (TGN) membrane proteins that contain acidic cluster sorting motifs, PACS-1 is also known to be involved in HIV-1 Nef-mediated removal of MHC-I from the cell surface to the TGN, thus enabling HIV-1 to escape immune surveillance. PACS-1 also controls the endosome to Golgi trafficking of furin and mannose-6-phosphate receptor by connecting the acidic-cluster-containing cytoplasmic domain of these molecules with the adapter-protein complex-1 (AP-1) of endosomal clathrin coated membrane pits. Usually seen in golgi apparatus, trans-Golgi network and localized in the paranuclear region, it is ubiquitously expressed in most of the tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACS1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene PACS1
Gene Accession No. Q6VY07, Q8K212
Gene Alias AI325977; cytosolic sorting protein PACS-1; KIAA1175; LOW QUALITY PROTEIN: phosphofurin acidic cluster sorting protein 1; phosphofurin acidic cluster sorting protein 1; MRD17; PACS1; PACS-1; Phosphofurin acidic cluster sorting protein 1
Gene Symbols PACS1
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human PACS-1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100052235, 107975, 483716, 514313, 55690
Target Species Human, Mouse, Canine, Monkey, Bovine, Horse, Marsupial
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.