Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PACS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543701
Description
Peptide sequence: AHQLPIAEAM LTYKQKSPDE ESSQKFIPFV GVVKVGIVEP SSATSGDSDD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 86%.
PACS2 is the multifunctional sorting protein that controls the endoplasmic reticulum (ER)-mitochondria communication, including the apposition of mitochondria with the ER and ER homeostasis. In addition, in response to apoptic inducer, PACS2 translocates BIB to mitochondria, which initiates a sequence of events including the formation of mitochondrial truncated BID, the release of cytochrome c, the activation of caspase-3 thereby causing cell death.PACS2 may also be involved in ion channel trafficking, directing acidic cluster-containing ion channels to distinct subcellular compartments.
Specifications
PACS2 | |
Polyclonal | |
Unconjugated | |
PACS2 | |
6720425G15Rik; AW208651; AW489976; E230011J22; FLJ25488; Kiaa0602; mKIAA0602; Pacs1l; PACS1-like protein; Pacs2; PACS-2; Phosphofurin acidic cluster sorting protein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23241 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86VP3 | |
PACS2 | |
Synthetic peptide directed towards the middle region of human PACS2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction