Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PACS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551575 Supplier Invitrogen™ Supplier No. PA551575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAWRDLFNKL EAQSAVQDTP DIVSRITQYI AGANCAHQLP IAEAMLTYKQ KSPDEESSQK FIPFVGVVKV GIVEPSSATS GDSDDAAPSG SGTLSSTPPS ASPAAKEASP TPPSSPSVSG G Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

PACS2 is the multifunctional sorting protein that controls the endoplasmic reticulum (ER)-mitochondria communication, including the apposition of mitochondria with the ER and ER homeostasis. In addition, in response to apoptic inducer, PACS2 translocates BIB to mitochondria, which initiates a sequence of events including the formation of mitochondrial truncated BID, the release of cytochrome c, the activation of caspase-3 thereby causing cell death.PACS2 may also be involved in ion channel trafficking, directing acidic cluster-containing ion channels to distinct subcellular compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PACS2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PACS2
Gene Accession No. Q86VP3
Gene Alias 6720425G15Rik; AW208651; AW489976; E230011J22; FLJ25488; Kiaa0602; mKIAA0602; Pacs1l; PACS1-like protein; Pacs2; PACS-2; Phosphofurin acidic cluster sorting protein 2
Gene Symbols PACS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PACS2. Recombinant protein control fragment (Product #RP-89005).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23241
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.