Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACSIN1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299230PCP
Description
PACSIN1 Polyclonal antibody specifically detects PACSIN1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)Specifications
PACSIN1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
KIAA1379SDPI, protein kinase C and casein kinase substrate in neurons 1, protein kinase C and casein kinase substrate in neurons protein 1, syndapin I | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PACSIN1. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
29993 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction