Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PACSIN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528852
Description
Recommended positive controls: Molt-4, mouse brain, rat brain. Predicted reactivity: Mouse (95%), Rat (95%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
PACSIN1 | |
Polyclonal | |
Unconjugated | |
Pacsin1 | |
A830061D09Rik; Dynamin PRD-interacting protein; dynamin proline-rich domain-interacting protein; H74; KIAA1379; mKIAA1379; Pacsin; Pacsin1; protein kinase C and casein kinase substrate in neurons 1; protein kinase C and casein kinase substrate in neurons protein 1; SdpI; synaptic, dynamin-associated protein I; syndapin; syndapin 1; syndapin I; syndapin-1; Syndapin-I | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23969, 29704, 29993 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q61644, Q9BY11, Q9Z0W5 | |
Pacsin1 | |
Full length human PACSIN1 Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction